siRNA Details
| Gene Symbol: CTAGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172241.2 | NP_758441.2 | 1 | 3206 | ||
| GenBank mRNA | AF177229.1 | - | 1 | 158 | ||
| AF273058.1 | - | 1 | 3213 | |||
| BC132871.1 | - | 1 | 3139 | 40146893 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTAGE1| Gene Symbol: CTAGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172241.2 | NP_758441.2 | 1 | 3206 | ||
| GenBank mRNA | AF177229.1 | - | 1 | 158 | ||
| AF273058.1 | - | 1 | 3213 | |||
| BC132871.1 | - | 1 | 3139 | 40146893 | ||