siRNA Details
| Gene Symbol: ZNF322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242797.1 | NP_001229726.1 | 5 | 2975 | ||
| NM_001242798.1 | NP_001229727.1 | 3 | 2830 | |||
| NM_001242799.1 | NP_001229728.1 | 3 | 2811 | |||
| NM_024639.4 | NP_078915.2 | 4 | 2900 | |||
| GenBank mRNA | AK027046.1 | - | 1 | 2220 | ||
| AY376736.1 | - | 4 | 2683 | |||

ZNF322
Genomic Map