siRNA Details
| Gene Symbol: OR10W1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207374.3 | NP_997257.2 | 1 | 577 | ||
| GenBank mRNA | AY358251.1 | - | 1 | 577 | ||
| BC132848.1 | - | 1 | 476 | 40146870 | ||
| BC132850.1 | - | 1 | 476 | 40146872 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR10W1| Gene Symbol: OR10W1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207374.3 | NP_997257.2 | 1 | 577 | ||
| GenBank mRNA | AY358251.1 | - | 1 | 577 | ||
| BC132848.1 | - | 1 | 476 | 40146870 | ||
| BC132850.1 | - | 1 | 476 | 40146872 | ||