siRNA Details
| Gene Symbol: KRTAP9-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031961.2 | NP_114167.2 | 1 | 765 | ||
| Gene Symbol: KRTAP9-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031963.2 | NP_114169.2 | 1 | 747 | ||

KRTAP9-2
Genomic Map