siRNA Details
| Gene Symbol: TSSK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032037.3 | NP_114426.1 | 1 | 1424 | ||
| GenBank mRNA | AF329483.1 | - | 1 | 1313 | ||
| AF348077.1 | - | 1 | 1449 | |||
| AK058166.1 | - | 2 | 1249 | |||
| AK172841.1 | - | 1 | 1794 | |||
| BC014611.2 | - | 1 | 1312 | 4826305 | ||

TSSK6
Genomic Map