siRNA Details
Gene Symbol: KRTAP4-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032524.1 | NP_115913.1 | 1 | 808 | ||
GenBank mRNA | AJ296168.1 | - | 2 | 631 | ||
AJ406936.1 | - | 1 | 808 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KRTAP4-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032524.1 | NP_115913.1 | 1 | 808 | ||
GenBank mRNA | AJ296168.1 | - | 2 | 631 | ||
AJ406936.1 | - | 1 | 808 |