siRNA Details
| Gene Symbol: KRTAP4-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032524.1 | NP_115913.1 | 1 | 624 | ||
| GenBank mRNA | AJ296168.1 | - | 2 | 447 | ||
| AJ406936.1 | - | 1 | 624 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP4-4| Gene Symbol: KRTAP4-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032524.1 | NP_115913.1 | 1 | 624 | ||
| GenBank mRNA | AJ296168.1 | - | 2 | 447 | ||
| AJ406936.1 | - | 1 | 624 | |||