siRNA Details
| Gene Symbol: PAGE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013435.2 | NP_001013453.1 | 2 | 131 | ||
| NM_130467.4 | NP_569734.2 | 2 | 330 | |||
| GenBank mRNA | AJ344352.1 | - | 2 | 300 | ||
| BC009230.2 | - | 2 | 319 | 3955765 | ||
| BC110803.1 | - | 2 | 65 | 3922538 | ||
| BP429046.1 | - | 2 | 135 | |||
| BQ437151.1 | - | 2 | 119 | |||

PAGE5
Genomic Map