siRNA Details
| Gene Symbol: DERL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002862.2 | NP_001002862.1 | 7 | 1034 | ||
| NM_198440.3 | NP_940842.2 | 6 | 1135 | |||
| GenBank mRNA | AK092582.1 | - | 1 | 179 | ||
| AK096976.1 | - | 5 | 1046 | |||
| AK125830.1 | - | 5 | 1336 | |||

DERL3
Genomic Map