siRNA Details
| Gene Symbol: ANKRD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152326.2 | NP_689539.1 | 4 | 1609 | ||
| XM_005267310.2 | XP_005267367.1 | 3 | 1746 | |||
| XM_005267311.3 | XP_005267368.1 | 3 | 1378 | |||
| GenBank mRNA | AK291868.1 | - | 3 | 1275 | ||
| BC015422.1 | - | 2 | 1408 | 4397794 | ||
| BC027479.1 | - | 2 | 1126 | 5114925 | ||
| BX161460.1 | - | 4 | 1609 | |||

ANKRD9
Genomic Map