siRNA Details
| Gene Symbol: OR2M5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004690.1 | NP_001004690.1 | 1 | 218 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2M5| Gene Symbol: OR2M5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004690.1 | NP_001004690.1 | 1 | 218 | ||