siRNA Details
| Gene Symbol: RIPPLY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009994.2 | NP_001009994.1 | 4 | 525 | ||
| NR_103525.1 | 3 | 431 | ||||
| GenBank mRNA | AK124105.1 | - | 1 | 3785 | ||
| BC130460.1 | - | 4 | 490 | 40146491 | ||
| BC132968.1 | - | 4 | 490 | 40146990 | ||
| BM782014.1 | - | 3 | 384 | |||

RIPPLY2
Genomic Map