siRNA Details
Gene Symbol: OR2A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001659.1 | NP_001001659.1 | 1 | 332 | ||
GenBank mRNA | BC136889.1 | - | 1 | 387 | 9020880 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR2A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001659.1 | NP_001001659.1 | 1 | 332 | ||
GenBank mRNA | BC136889.1 | - | 1 | 387 | 9020880 |