siRNA Details
| Gene Symbol: TLDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304783.1 | NP_001291712.1 | 6 | 966 | ||
| NM_080628.2 | NP_542195.1 | 7 | 1062 | |||
| GenBank mRNA | AK123544.1 | - | 7 | 1016 | ||
| BG163960.1 | - | 560 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TLDC2| Gene Symbol: TLDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304783.1 | NP_001291712.1 | 6 | 966 | ||
| NM_080628.2 | NP_542195.1 | 7 | 1062 | |||
| GenBank mRNA | AK123544.1 | - | 7 | 1016 | ||
| BG163960.1 | - | 560 | ||||