siRNA Details
| Gene Symbol: LYRM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001660.2 | NP_001001660.2 | 3 | 675 | ||
| XM_005253319.4 | XP_005253376.1 | 3 | 678 | |||
| XM_005253320.4 | XP_005253377.1 | 4 | 782 | |||
| XM_017018850.1 | XP_016874339.1 | 3 | 2096 | |||
| GenBank mRNA | AK057730.1 | - | 3 | 679 | ||
| AK095164.1 | - | 2 | 2176 | |||
| BC071769.1 | - | 3 | 609 | 4246777 | ||
| BC100276.1 | - | 3 | 626 | 6178563 | ||

LYRM5
Genomic Map