siRNA Details
Gene Symbol: SSTR5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027242.1 | 4 | 1565 | |||
GenBank mRNA | AK056814.1 | - | 4 | 1509 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SSTR5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027242.1 | 4 | 1565 | |||
GenBank mRNA | AK056814.1 | - | 4 | 1509 |