siRNA Details
| Gene Symbol: SZT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015284.3 | NP_056099.3 | 4 | 554 | ||
| XM_005270686.3 | XP_005270743.1 | 5 | 615 | |||
| XM_006710501.3 | XP_006710564.1 | 4 | 502 | |||
| XM_011541106.2 | XP_011539408.1 | 4 | 653 | |||
| XM_017000819.1 | XP_016856308.1 | 5 | 615 | |||
| XM_017000821.1 | XP_016856310.1 | 1 | 119 | |||
| GenBank mRNA | AK091821.1 | - | 4 | 475 | ||
| BC017576.1 | - | 4 | 552 | |||
| BC051343.1 | - | 4 | 488 | |||
| BU101724.1 | - | 1 | 47 | |||

SZT2
Genomic Map