siRNA Details
| Gene Symbol: C22orf42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010859.1 | NP_001010859.1 | 7 | 650 | ||
| XM_011529922.2 | XP_011528224.1 | 8 | 1149 | |||
| XM_017028629.1 | XP_016884118.1 | 8 | 683 | |||
| XM_017028630.1 | XP_016884119.1 | 6 | 1059 | |||
| GenBank mRNA | CR456485.1 | - | 7 | 650 | ||

C22orf42
Genomic Map