siRNA Details
| Gene Symbol: OR6B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173351.1 | NP_775486.1 | 1 | 961 | ||
| XM_011510671.2 | XP_011508973.1 | 3 | 1358 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR6B3| Gene Symbol: OR6B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173351.1 | NP_775486.1 | 1 | 961 | ||
| XM_011510671.2 | XP_011508973.1 | 3 | 1358 | |||