siRNA Details
| Gene Symbol: FAM53A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013622.3 | NP_001013644.1 | 4, 5 | 1084 | ||
| NM_001174070.2 | NP_001167541.1 | 4, 5 | 1131 | |||
| XM_005247948.3 | XP_005248005.1 | 4, 5 | 1781 | |||
| XM_011513402.2 | XP_011511704.1 | 4, 5 | 1787 | |||
| GenBank mRNA | AK125566.1 | - | 4, 5 | 1079 | ||
| BC070112.1 | - | 4, 5 | 1084 | 5268061 | ||

FAM53A
Genomic Map