siRNA Details
| Gene Symbol: C8orf48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007090.2 | NP_001007091.1 | 1 | 433 | ||
| GenBank mRNA | AF394908.1 | - | 1 | 286 | ||
| AK058131.1 | - | 1 | 433 | |||
| BC031245.1 | - | 1 | 381 | 5266320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C8orf48| Gene Symbol: C8orf48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007090.2 | NP_001007091.1 | 1 | 433 | ||
| GenBank mRNA | AF394908.1 | - | 1 | 286 | ||
| AK058131.1 | - | 1 | 433 | |||
| BC031245.1 | - | 1 | 381 | 5266320 | ||