siRNA Details
| Gene Symbol: SPIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027139.1 | 3 | 598 | |||
| GenBank mRNA | AK123122.1 | - | 3 | 662 | ||
| BC018073.1 | - | 3 | 542 | |||
| BC032490.1 | - | 3 | 552 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPIN3| Gene Symbol: SPIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027139.1 | 3 | 598 | |||
| GenBank mRNA | AK123122.1 | - | 3 | 662 | ||
| BC018073.1 | - | 3 | 542 | |||
| BC032490.1 | - | 3 | 552 | |||