siRNA Details
| Gene Symbol: ZSWIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK091621.1 | - | 1 | 604 | ||
| BC137249.1 | - | 1 | 526 | |||
| BC137250.1 | - | 1 | 526 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZSWIM8| Gene Symbol: ZSWIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK091621.1 | - | 1 | 604 | ||
| BC137249.1 | - | 1 | 526 | |||
| BC137250.1 | - | 1 | 526 | |||