siRNA Details
| Gene Symbol: LINC01599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131171.1 | 2 | 112 | |||
| GenBank mRNA | AF390030.1 | - | 3 | 437 | ||
| DA935043.1 | - | 2 | 114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01599| Gene Symbol: LINC01599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131171.1 | 2 | 112 | |||
| GenBank mRNA | AF390030.1 | - | 3 | 437 | ||
| DA935043.1 | - | 2 | 114 | |||