siRNA Details
| Gene Symbol: TMEM201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010866.3 | NP_001010866.1 | 6 | 3110 | ||
| GenBank mRNA | BC121187.1 | - | 6 | 3065 | 3870153 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM201| Gene Symbol: TMEM201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010866.3 | NP_001010866.1 | 6 | 3110 | ||
| GenBank mRNA | BC121187.1 | - | 6 | 3065 | 3870153 | |