siRNA Details
| Gene Symbol: EPGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270989.1 | NP_001257918.1 | 4 | 353 | ||
| XM_005265666.4 | XP_005265723.1 | 4 | 1225 | |||
| XM_011531817.2 | XP_011530119.1 | 4 | 1211 | |||
| GenBank mRNA | AK289455.1 | - | 4 | 351 | ||
| AY358920.1 | - | 4 | 309 | |||
| CB150835.1 | - | 4 | 326 | |||
| DQ235269.1 | - | 4 | 337 | |||

EPGN
Genomic Map