siRNA Details
| Gene Symbol: TMEM211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001663.1 | NP_001001663.1 | 3 | 447 | ||
| XM_011530094.2 | XP_011528396.1 | 2 | 1324 | |||
| GenBank mRNA | CR456388.1 | - | 3 | 447 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM211| Gene Symbol: TMEM211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001663.1 | NP_001001663.1 | 3 | 447 | ||
| XM_011530094.2 | XP_011528396.1 | 2 | 1324 | |||
| GenBank mRNA | CR456388.1 | - | 3 | 447 | ||