siRNA Details
| Gene Symbol: CFAP126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013625.3 | NP_001013647.2 | 3 | 132 | ||
| GenBank mRNA | DT215043.1 | - | 3 | 132 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CFAP126| Gene Symbol: CFAP126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013625.3 | NP_001013647.2 | 3 | 132 | ||
| GenBank mRNA | DT215043.1 | - | 3 | 132 | ||