siRNA Details
Gene Symbol: LOC284912 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC001801.1 | - | 2 | 683 | 3355513 | |
BG471420.1 | - | 2 | 682 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC284912 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC001801.1 | - | 2 | 683 | 3355513 | |
BG471420.1 | - | 2 | 682 |