siRNA Details
| Gene Symbol: KRTAP13-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181621.3 | NP_853652.1 | 1 | 697 | ||
| GenBank mRNA | BX104478.1 | - | 1 | 714 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP13-2| Gene Symbol: KRTAP13-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181621.3 | NP_853652.1 | 1 | 697 | ||
| GenBank mRNA | BX104478.1 | - | 1 | 714 | ||