siRNA Details
| Gene Symbol: KRTAP19-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181609.3 | NP_853640.1 | 1 | 382 | ||
| GenBank mRNA | AF139541.1 | - | 1 | 382 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP19-3| Gene Symbol: KRTAP19-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181609.3 | NP_853640.1 | 1 | 382 | ||
| GenBank mRNA | AF139541.1 | - | 1 | 382 | ||