siRNA Details
| Gene Symbol: LRRC37A11P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033753.2 | 4 | 3043 | |||
| GenBank mRNA | AK125814.1 | - | 4 | 3042 | ||
| BC111725.1 | - | 4 | 2769 | 40028479 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC37A11P| Gene Symbol: LRRC37A11P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033753.2 | 4 | 3043 | |||
| GenBank mRNA | AK125814.1 | - | 4 | 3042 | ||
| BC111725.1 | - | 4 | 2769 | 40028479 | ||