siRNA Details
| Gene Symbol: LCE1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276331.1 | NP_001263260.1 | 3 | 450 | ||
| NM_178351.3 | NP_848128.1 | 2 | 540 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LCE1C| Gene Symbol: LCE1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276331.1 | NP_001263260.1 | 3 | 450 | ||
| NM_178351.3 | NP_848128.1 | 2 | 540 | |||