siRNA Details
| Gene Symbol: CCDC172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198515.2 | NP_940917.1 | 3, 4 | 412 | ||
| XM_011539771.2 | XP_011538073.1 | 3, 4 | 740 | |||
| XM_017016206.1 | XP_016871695.1 | 3, 4 | 330 | |||
| GenBank mRNA | AJ457798.1 | - | 2, 3 | 394 | ||
| BC044830.1 | - | 3, 4 | 412 | 5166167 | ||

CCDC172
Genomic Map