siRNA Details
Gene Symbol: TMEM110-MUSTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198974.2 | NP_001185903.2 | 3 | 371 |
Gene Symbol: TMEM110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198563.2 | NP_940965.1 | 3 | 371 | ||
GenBank mRNA | AY726591.1 | - | 3 | 135 | ||
BC047015.1 | - | 3 | 321 | 5199769 | ||
BX360795.2 | - | 3 | 386 |
