siRNA Details
| Gene Symbol: SLC22A25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199352.3 | NP_955384.3 | 1 | 92 | ||
| XM_017017684.1 | XP_016873173.1 | 4 | 1171 | |||
| XM_017017685.1 | XP_016873174.1 | 5 | 1299 | |||
| XM_017017686.1 | XP_016873175.1 | 4 | 1607 | |||
| XM_017017687.1 | XP_016873176.1 | 4 | 1712 | |||
| XM_017017695.1 | XP_016873184.1 | 4 | 1712 | |||
| GenBank mRNA | AY437532.1 | - | 1 | 92 | ||
| BC101315.1 | - | 1 | 87 | |||
| BC101316.1 | - | 1 | 92 | |||
| BC101317.1 | - | 1 | 92 | |||

SLC22A25
Genomic Map