siRNA Details
| Gene Symbol: OR5M10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004741.1 | NP_001004741.1 | 1 | 472 | ||
| GenBank mRNA | BC140733.1 | - | 1 | 538 | 9021601 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5M10| Gene Symbol: OR5M10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004741.1 | NP_001004741.1 | 1 | 472 | ||
| GenBank mRNA | BC140733.1 | - | 1 | 538 | 9021601 | |