siRNA Details
| Gene Symbol: SPRYD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127482.2 | NP_001120954.1 | 2, 3 | 536 | ||
| NM_020456.3 | NP_065189.1 | 3, 4 | 653 | |||
| XM_017020677.1 | XP_016876166.1 | 3, 4 | 653 | |||
| GenBank mRNA | AF052154.1 | - | 3, 4 | 452 | ||
| AF055016.1 | - | 3, 4 | 452 | |||
| AF334405.1 | - | 3, 4 | 492 | |||
| AK290576.1 | - | 2, 3 | 375 | |||
| AK308818.1 | - | 3, 4 | 486 | |||
| AK316576.1 | - | 3, 4 | 459 | |||
| BC022519.1 | - | 2, 3 | 342 | 4792366 | ||

SPRYD7
Genomic Map