siRNA Details
| Gene Symbol: OR2M7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004691.1 | NP_001004691.1 | 1 | 893 | ||
| GenBank mRNA | BC136944.1 | - | 1 | 922 | 9020936 | |
| BC136946.1 | - | 1 | 922 | 9020938 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2M7| Gene Symbol: OR2M7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004691.1 | NP_001004691.1 | 1 | 893 | ||
| GenBank mRNA | BC136944.1 | - | 1 | 922 | 9020936 | |
| BC136946.1 | - | 1 | 922 | 9020938 | ||