siRNA Details
| Gene Symbol: METTL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304467.1 | NP_001291396.1 | 7 | 2216 | ||
| NM_001304468.1 | NP_001291397.1 | 6 | 1957 | |||
| NM_212554.3 | NP_997719.2 | 7 | 2212 | |||
| GenBank mRNA | AK022354.1 | - | 3 | 1663 | ||
| AL832292.1 | - | 7 | 2113 | |||

METTL10
Genomic Map