siRNA Details
| Gene Symbol: FAR2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026758.2 | 12 | 3363 | |||
| GenBank mRNA | AK125921.1 | - | 12 | 3364 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAR2P1| Gene Symbol: FAR2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026758.2 | 12 | 3363 | |||
| GenBank mRNA | AK125921.1 | - | 12 | 3364 | ||