siRNA Details
| Gene Symbol: RNF165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256758.1 | NP_001243687.1 | 4 | 373 | ||
| NM_152470.2 | NP_689683.2 | 6 | 797 | |||
| GenBank mRNA | AK122819.1 | - | 4 | 373 | ||
| AK127467.1 | - | 6 | 797 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF165| Gene Symbol: RNF165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256758.1 | NP_001243687.1 | 4 | 373 | ||
| NM_152470.2 | NP_689683.2 | 6 | 797 | |||
| GenBank mRNA | AK122819.1 | - | 4 | 373 | ||
| AK127467.1 | - | 6 | 797 | |||