siRNA Details
| Gene Symbol: RNF165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256758.1 | NP_001243687.1 | 6 | 3451 | ||
| NM_152470.2 | NP_689683.2 | 8 | 3875 | |||
| XM_011526016.2 | XP_011524318.1 | 7 | 3779 | |||
| GenBank mRNA | AK096648.1 | - | 1 | 1688 | ||
| AK122819.1 | - | 6 | 3451 | |||
| AK123883.1 | - | 2 | 2961 | |||
| AK127467.1 | - | 8 | 3875 | |||

RNF165
Genomic Map