siRNA Details
| Gene Symbol: FLJ25758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024372.1 | 1 | 996 | |||
| GenBank mRNA | AK098624.1 | - | 1 | 1003 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ25758| Gene Symbol: FLJ25758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024372.1 | 1 | 996 | |||
| GenBank mRNA | AK098624.1 | - | 1 | 1003 | ||