siRNA Details
| Gene Symbol: TCL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028288.1 | 5 | 1485 | |||
| GenBank mRNA | AB035332.1 | - | 2 | 3264 | ||
| AB035333.1 | - | 2 | 3264 | |||
| AB035334.1 | - | 3 | 2296 | |||
| AB035335.1 | - | 2 | 1841 | |||
| AB035337.1 | - | 5 | 1487 | |||
| AB035338.1 | - | 5 | 1682 | |||
| AF195821.1 | - | 1 | 87 | |||
| BC041075.1 | - | 2 | 1655 | 5923478 | ||
| BX161462.1 | - | 7 | 1530 | |||

TCL6
Genomic Map