siRNA Details
| Gene Symbol: ZNF275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC001545.1 | - | 1 | 835 | ||
| BC136634.1 | - | 1 | 476 | 9020624 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF275| Gene Symbol: ZNF275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC001545.1 | - | 1 | 835 | ||
| BC136634.1 | - | 1 | 476 | 9020624 | ||