siRNA Details
| Gene Symbol: JPH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020655.3 | NP_065706.2 | 2 | 1267 | ||
| NR_073379.2 | 2 | 721 | ||||
| GenBank mRNA | AB042636.1 | - | 2 | 1176 | ||
| AK055486.1 | - | 1 | 78 | |||
| AK295518.1 | - | 2 | 721 | |||
| BC036533.2 | - | 2 | 1249 | 5264853 | ||

JPH3
Genomic Map