siRNA Details
| Gene Symbol: TTYH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005367.2 | NP_001005367.1 | 4 | 733 | ||
| NM_001201461.1 | NP_001188390.1 | 4 | 733 | |||
| NM_020659.3 | NP_065710.1 | 4 | 733 | |||
| XM_017027025.1 | XP_016882514.1 | 4 | 577 | |||
| XM_017027026.1 | XP_016882515.1 | 4 | 577 | |||
| GenBank mRNA | AB162930.1 | - | 4 | 616 | ||
| AB188496.1 | - | 4 | 733 | |||
| AF177909.1 | - | 4 | 647 | |||
| AK126690.1 | - | 3 | 644 | |||
| AK289468.1 | - | 4 | 696 | |||
| AK293468.1 | - | 6 | 820 | |||
| BC011347.1 | - | 4 | 638 | 4215136 | ||
| BX402889.2 | - | 4 | 713 | |||

TTYH1
Genomic Map