siRNA Details
| Gene Symbol: FOXO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170931.1 | NP_001164402.1 | 2, 3 | 668 | ||
| NM_005938.3 | NP_005929.2 | 1, 2 | 833 | |||
| GenBank mRNA | AF384029.1 | - | 2, 3 | 288 | ||
| AK300142.1 | - | 1, 2 | 779 | |||
| BC106761.2 | - | 1, 2 | 559 | 40025381 | ||
| X93996.1 | - | 1, 2 | 623 | |||

FOXO4
Genomic Map