siRNA Details
| Gene Symbol: Olr1690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000274.1 | NP_001000274.1 | 1 | 603 | ||
| Gene Symbol: LOC100910696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772684.1 | XP_008770906.1 | 626 | |||

Olr1690
Genomic Map